Learn More
Cayman Chemical (±)-Norepinephrine (hydrochloride), Purity: ≥95%, CAS: 55-27-6, 500 mg
SDP
Catalog No. 502913221
Encompass_Preferred
A racemic mixture of of (–)-norepinephrine and (+)-norepinephrine; induces cAMP accumulation in rat cerebral cortical membranes at 1-100 µM; induces contraction of bovine anterior cerebral, middle cerebral, and internal carotid arterial strips (EC50s = 0.91, 0.92, and 0.87 µM, respectively) but induces relaxation of bovine posterior cerebral arterial strips (EC50 = 0.95 µM)
Catalog No. 50-291-3221 Supplier Cayman Chemical Supplier No. 35580500 MG
A racemic mixture of of (–)-norepinephrine and (+)-norepinephrine; induces cAMP accumulation in rat cerebral cortical membranes at 1-100 µM; induces contraction of bovine anterior cerebral, middle cerebral, and internal carotid arterial strips (EC50s = 0.91, 0.92, and 0.87 µM, respectively) but induces relaxation of bovine posterior cerebral arterial strips (EC50 = 0.95 µM)