Learn More
Medchemexpress LLC HY-B1710 100mg Medchemexpress, Norethindrone acetate CAS:51-98-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1710100MG
Encompass_Preferred
Medchemexpress, HY-B1710 100mg Norethindrone acetate CAS:51-98-9 Norethindrone acetate is a female progestin approved by FDA for the treatment of endometriosis, uterine bleeding caused by abnormal hormone levels, and secondary amenorrhea. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2992