Learn More
Selleck Chemical LLC Norethindrone acetate
SDP
Catalog No. 5000003294
Encompass_Preferred
Norethindrone acetate (19-Norethindrone acetate NA) is a common synthetic hormone used in oral contraceptives Norethindrone acetate is a well-tolerated effective option to manage pain and bleeding for all stages of endometriosis
Catalog No. 50-000-03294 Supplier Selleck Chemical LLC Supplier No. S578525MG
Norethindrone acetate (19-Norethindrone acetate NA) is a common synthetic hormone used in oral contraceptives Norethindrone acetate is a well-tolerated effective option to manage pain and bleeding for all stages of endometriosis