Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Norethindrone acetate | 51-98-9 | MFCD00271615 | 99.5% | 340.46 g·mol⁻¹ | C22H28O3 | 10 MG
SDP

Catalog No. 5000298902
Encompass_Preferred

Norethindrone acetate is provided as an analytical standard for research and analytical applications; it is a high-purity synthetic progestin supplied as a small, solid reference material for assay development and chromatographic analysis.

  • Analytical standard for assay development and chromatography.
  • High purity: 99.47%.
  • Chemical formula: C22H28O3.
  • Molecular weight: 340.46 g·mol⁻¹.
  • Packaged as a 10 MG vial for laboratory use.

Catalog No. 50-002-98902 Supplier Medchemexpress LLC Supplier No. HYB1710R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Norethindrone acetate is provided as an analytical standard for research and analytical applications; it is a high-purity synthetic progestin supplied as a small, solid reference material for assay development and chromatographic analysis.

  • Analytical standard for assay development and chromatography.
  • High purity: 99.47%.
  • Chemical formula: C22H28O3.
  • Molecular weight: 340.46 g·mol⁻¹.
  • Packaged as a 10 MG vial for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.