Learn More
Medchemexpress LLC Norethindrone-d6 | 2376036-05-2 | 99.9% | 1 MG
SDP
Catalog No. 5000407844
Encompass_Preferred
Norethindrone-d6 is a deuterium-labeled version of Norethindrone, a female progestin approved by the FDA for treating endometriosis, uterine bleeding caused by abnormal hormone levels, and secondary amenorrhea. It is also a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules can act as tracers for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07844 Supplier Medchemexpress LLC Supplier No. HYB0554S1MG
Norethindrone-d6 is a deuterium-labeled version of Norethindrone, a female progestin approved by the FDA for treating endometriosis, uterine bleeding caused by abnormal hormone levels, and secondary amenorrhea. It is also a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules can act as tracers for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.