Learn More
Medchemexpress LLC Norethindrone-d6 | 2376036-05-2 | 99.9% | 5 MG
SDP
Catalog No. 5000408348
Encompass_Preferred
Norethindrone-d6 is the deuterium-labeled version of Norethindrone, a female progestin approved by the FDA for treating endometriosis, uterine bleeding due to abnormal hormone levels, and secondary amenorrhea. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Can be used as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation as tracers during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
  • Its Alkyne group enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
Catalog No. 50-004-08348 Supplier Medchemexpress LLC Supplier No. HYB0554S5MG
Norethindrone-d6 is the deuterium-labeled version of Norethindrone, a female progestin approved by the FDA for treating endometriosis, uterine bleeding due to abnormal hormone levels, and secondary amenorrhea. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Can be used as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation as tracers during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
  • Its Alkyne group enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.