Learn More
Medchemexpress LLC Norethisterone enanthate | 3836-23-5 | 99.19% | C27H38O3 | 250 MG
SDP
Catalog No. 5000358526
Encompass_Preferred
Norethisterone enanthate is a long-acting parenteral progestogen, orally active, and functions as a click chemistry reagent. It contains an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This compound is intended for research use only.
  • Long-acting parenteral progestogen
  • Orally active
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can be used as an intramuscular contraceptive agent in vitro
  • Inhibits fertility in female rats in vivo
  • White to off-white solid appearance
  • Soluble in DMSO (200 mg/mL)
Catalog No. 50-003-58526 Supplier Medchemexpress LLC Supplier No. 250MGHYA0285
Norethisterone enanthate is a long-acting parenteral progestogen, orally active, and functions as a click chemistry reagent. It contains an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This compound is intended for research use only.
  • Long-acting parenteral progestogen
  • Orally active
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can be used as an intramuscular contraceptive agent in vitro
  • Inhibits fertility in female rats in vivo
  • White to off-white solid appearance
  • Soluble in DMSO (200 mg/mL)