Learn More
Medchemexpress LLC Norethisterone enanthate | 3836-23-5 | 99.5% | C27H38O3 | 1 ML
SDP
Catalog No. 5000432211
Encompass_Preferred
Norethisterone enanthate is a long-acting parenteral progestogen used as an intramuscular contraceptive agent. It is also an orally active click chemistry reagent containing an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Long-acting parenteral progestogen
  • Functions as an intramuscular contraceptive agent
  • Orally active
  • Contains an alkyne group for click chemistry applications
  • Powder stable at -20°C for 3 years or 4°C for 2 years
  • Solution stable at -80°C for 2 years or -20°C for 1 year
  • Store stock solutions aliquoted to prevent inactivation from repeated freeze-thaw cycles
Catalog No. 50-004-32211 Supplier Medchemexpress LLC Supplier No. HYA028510MM/1ML
Norethisterone enanthate is a long-acting parenteral progestogen used as an intramuscular contraceptive agent. It is also an orally active click chemistry reagent containing an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Long-acting parenteral progestogen
  • Functions as an intramuscular contraceptive agent
  • Orally active
  • Contains an alkyne group for click chemistry applications
  • Powder stable at -20°C for 3 years or 4°C for 2 years
  • Solution stable at -80°C for 2 years or -20°C for 1 year
  • Store stock solutions aliquoted to prevent inactivation from repeated freeze-thaw cycles