Learn More
Medchemexpress LLC Norethisterone enanthate | 3836-23-5 | 99.5% | C27H38O3 | 100 MG
SDP
Catalog No. 5000433510
Encompass_Preferred
Norethisterone enanthate is a long-acting parenteral progestogen that is orally active. It is also described as a click chemistry reagent, containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups. It can be used as an intramuscular contraceptive agent.
  • Long-acting parenteral progestogen
  • Orally active
  • Functions as a click chemistry reagent with an alkyne group
  • Can participate in CuAAc with azide-containing molecules
  • Has been used as an intramuscular contraceptive agent
Catalog No. 50-004-33510 Supplier Medchemexpress LLC Supplier No. HYA0285100MG
Norethisterone enanthate is a long-acting parenteral progestogen that is orally active. It is also described as a click chemistry reagent, containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups. It can be used as an intramuscular contraceptive agent.
  • Long-acting parenteral progestogen
  • Orally active
  • Functions as a click chemistry reagent with an alkyne group
  • Can participate in CuAAc with azide-containing molecules
  • Has been used as an intramuscular contraceptive agent