Learn More
Medchemexpress LLC Norfenefrine (hydrochloride) | 4779-94-6 | 98.9% | 250 MG
SDP
Catalog No. 5000414335
Encompass_Preferred
Norfenefrine hydrochloride is an orally active, endogenously found α-adrenergic agonist. It is suitable for research on female stress incontinence.
  • Orally active
  • Endogenously found α-adrenergic agonist
  • Can be used for research of female stress incontinence
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-14335 Supplier Medchemexpress LLC Supplier No. 250MGHYB1711A
Norfenefrine hydrochloride is an orally active, endogenously found α-adrenergic agonist. It is suitable for research on female stress incontinence.
  • Orally active
  • Endogenously found α-adrenergic agonist
  • Can be used for research of female stress incontinence
  • Solid appearance
  • White to off-white color