Learn More
Medchemexpress LLC (+)-Norfenfluramine hydrochloride | 37936-89-3 | MFCD11045299 | 99.8% | 239.67 g/mol | C10H13ClF3N | 25 MG
SDP
Catalog No. 5000307724
Encompass_Preferred
(+)-Norfenfluramine hydrochloride is the optically active hydrochloride salt of norfenfluramine supplied as a research-grade reagent. It is a major hepatic metabolite of fenfluramine and a selective 5-HT2B receptor agonist used in in vitro pharmacology, receptor binding, and metabolic studies. Available in a small, high-purity pack suitable for analytical and experimental use.
  • High purity suitable for analytical and pharmacological studies.
  • Hydrochloride salt form for improved solubility in polar solvents.
  • Optically active (+) enantiomer for stereoselective research.
  • Documented activity as a 5-HT2B receptor agonist for receptor studies.
  • Small pack size supports limited-scale experiments and reference standards.
Catalog No. 50-003-07724 Supplier Medchemexpress LLC Supplier No. HY118010A25MG
(+)-Norfenfluramine hydrochloride is the optically active hydrochloride salt of norfenfluramine supplied as a research-grade reagent. It is a major hepatic metabolite of fenfluramine and a selective 5-HT2B receptor agonist used in in vitro pharmacology, receptor binding, and metabolic studies. Available in a small, high-purity pack suitable for analytical and experimental use.
  • High purity suitable for analytical and pharmacological studies.
  • Hydrochloride salt form for improved solubility in polar solvents.
  • Optically active (+) enantiomer for stereoselective research.
  • Documented activity as a 5-HT2B receptor agonist for receptor studies.
  • Small pack size supports limited-scale experiments and reference standards.