Learn More
Med Vet International Norfenicol (Florfenicol) inj 300mg/mL, 100mL
Catalog No. 502827767
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Cattle: Treatment of respiratory tract infections in clinically diseased cattle due to Mannheimia haemolytica, Pasteurella multocida and Histophilus somni, susceptible to Florfenicol Swine: Treatment of acute outbreaks of respiratory disease caused by strains of Actinobacillus pleuropneumoniae and Pasteurella multocida, susceptible to Florfenicol.
Catalog No. 50-282-7767 Supplier Med Vet International Supplier No. RXNORFENICOL100
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Cattle: Treatment of respiratory tract infections in clinically diseased cattle due to Mannheimia haemolytica, Pasteurella multocida and Histophilus somni, susceptible to Florfenicol Swine: Treatment of acute outbreaks of respiratory disease caused by strains of Actinobacillus pleuropneumoniae and Pasteurella multocida, susceptible to Florfenicol.