Learn More
Medchemexpress LLC Norfloxacin | 70458-96-7 | 99.8% | 250 MG
SDP
Catalog No. 5000406488
Encompass_Preferred
Norfloxacin (Standard) is the analytical standard of Norfloxacin. This product is intended for research and analytical applications. Norfloxacin is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria, functioning by inhibiting DNA gyrase. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Broad-spectrum antibiotic
  • Active against Gram-positive and Gram-negative bacteria
  • Inhibits DNA gyrase
  • Suitable for research and analytical applications
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-06488 Supplier Medchemexpress LLC Supplier No. 250MGHYB0132R
Norfloxacin (Standard) is the analytical standard of Norfloxacin. This product is intended for research and analytical applications. Norfloxacin is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria, functioning by inhibiting DNA gyrase. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Broad-spectrum antibiotic
  • Active against Gram-positive and Gram-negative bacteria
  • Inhibits DNA gyrase
  • Suitable for research and analytical applications
  • Used in HPLC, GC, and MS research experiments