Learn More
Medchemexpress LLC Norfloxacin | 70458-96-7 | 99.8% | 50 MG
SDP
Catalog No. 5000450854
Encompass_Preferred
Norfloxacin (Standard) is the analytical standard of Norfloxacin, intended for research and analytical applications. Norfloxacin (MK-0366) is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria, which functions by inhibiting DNA gyrase. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications
  • Broad-spectrum antibiotic
  • Active against Gram-positive and Gram-negative bacteria
  • Inhibits DNA gyrase
  • Used in HPLC, GC, and MS experiments
  • White to off-white solid appearance
Catalog No. 50-004-50854 Supplier Medchemexpress LLC Supplier No. HYB0132R50MG
Norfloxacin (Standard) is the analytical standard of Norfloxacin, intended for research and analytical applications. Norfloxacin (MK-0366) is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria, which functions by inhibiting DNA gyrase. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications
  • Broad-spectrum antibiotic
  • Active against Gram-positive and Gram-negative bacteria
  • Inhibits DNA gyrase
  • Used in HPLC, GC, and MS experiments
  • White to off-white solid appearance