Learn More
Medchemexpress LLC Norfloxacin | 70458-96-7 | MFCD00079532 | 98.3% | 319.33 | 500 MG
SDP
Catalog No. 5000365918
Encompass_Preferred
Norfloxacin (MK-0366) is a broad-spectrum antibiotic active against both Gram-positive and Gram-negative bacteria, functioning by inhibiting DNA gyrase. It is a synthetic chemotherapeutic antibacterial agent sometimes used for common and complicated urinary tract infections.
  • Broad-spectrum antibiotic
  • Active against Gram-positive bacteria
  • Active against Gram-negative bacteria
  • Inhibits DNA gyrase
  • Synthetic chemotherapeutic antibacterial agent
  • Used for urinary tract infections
Catalog No. 50-003-65918 Supplier Medchemexpress LLC Supplier No. 500MGHYB0132
Norfloxacin (MK-0366) is a broad-spectrum antibiotic active against both Gram-positive and Gram-negative bacteria, functioning by inhibiting DNA gyrase. It is a synthetic chemotherapeutic antibacterial agent sometimes used for common and complicated urinary tract infections.
  • Broad-spectrum antibiotic
  • Active against Gram-positive bacteria
  • Active against Gram-negative bacteria
  • Inhibits DNA gyrase
  • Synthetic chemotherapeutic antibacterial agent
  • Used for urinary tract infections