Learn More
Abcam Norflo x acin, Antibacterial agent, 5G
Catalog No. 502706787
Encompass_Preferred
MW 319.33 Da, Purity >99%. Broad spectrum antibacterial agent active against Gram-positive and Gram-negative bacteria. Inhibits cell division by inhibiting DNA gyrase. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6787 Supplier Abcam Supplier No. AB1412195G
MW 319.33 Da, Purity >99%. Broad spectrum antibacterial agent active against Gram-positive and Gram-negative bacteria. Inhibits cell division by inhibiting DNA gyrase. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement