Learn More
Apexbio Technology LLC Norfloxacin hydrochloride 104142-93-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088588
Encompass_Preferred
Norfloxacin hydrochloride is a small-molecule inhibitor targeting bacterial DNA gyrase It is designed to inhibit DNA gyrase an enzyme essential for the supercoiling and replication of bacterial DNA thereby disrupting bacterial DNA synthesis and interfering with bacterial replication and growth Norfloxacin hydrochloride exerts its biological activity primarily through the inhibition of DNA gyrase by binding within the enzyme s active site impeding its catalytic activity Based on these pharmacological properties Norfloxacin hydrochloride holds research potential in the study of bacterial resistance mechanisms bacterial enzyme inhibition assays antimicrobial susceptibility testing and therapeutic research on antibacterial treatment strategies against Gram-positive and Gram-negative bacteria
Catalog No. 50-000-88588 Supplier Apexbio Technology LLC Supplier No. B12215.1
Norfloxacin hydrochloride is a small-molecule inhibitor targeting bacterial DNA gyrase It is designed to inhibit DNA gyrase an enzyme essential for the supercoiling and replication of bacterial DNA thereby disrupting bacterial DNA synthesis and interfering with bacterial replication and growth Norfloxacin hydrochloride exerts its biological activity primarily through the inhibition of DNA gyrase by binding within the enzyme s active site impeding its catalytic activity Based on these pharmacological properties Norfloxacin hydrochloride holds research potential in the study of bacterial resistance mechanisms bacterial enzyme inhibition assays antimicrobial susceptibility testing and therapeutic research on antibacterial treatment strategies against Gram-positive and Gram-negative bacteria