Learn More
Medchemexpress LLC Norfluoxetine-d5 hydrochloride | 1188265-34-0 | 99.3% | 336.79 | 10 MG
SDP
Catalog No. 5000452102
Encompass_Preferred
Norfluoxetine-d5 hydrochloride is a deuterium labeled chemical compound. It is a stable heavy isotope of hydrogen, carbon, and other elements that have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-52102 Supplier Medchemexpress LLC Supplier No. HYW011235S10MG
Add to Cart
Add to Cart
Norfluoxetine-d5 hydrochloride is a deuterium labeled chemical compound. It is a stable heavy isotope of hydrogen, carbon, and other elements that have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs