Learn More
Medchemexpress LLC Norfluoxetine-d5 hydrochloride | 1188265-34-0 | 99.3% | 336.79 | 5 MG
SDP
Catalog No. 5000453620
Encompass_Preferred
Norfluoxetine-d5 (hydrochloride) is a deuterium labeled compound, specifically 3-Phenyl-3-(4-(trifluoromethyl)phenoxy)propan-1-amine hydrochloride. Stable heavy isotopes of elements such as hydrogen and carbon are integrated into drug molecules, primarily to serve as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be utilized as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Deuteration can potentially influence the pharmacokinetic and metabolic characteristics of drugs.
Catalog No. 50-004-53620 Supplier Medchemexpress LLC Supplier No. HYW011235S5MG
Add to Cart
Add to Cart
Norfluoxetine-d5 (hydrochloride) is a deuterium labeled compound, specifically 3-Phenyl-3-(4-(trifluoromethyl)phenoxy)propan-1-amine hydrochloride. Stable heavy isotopes of elements such as hydrogen and carbon are integrated into drug molecules, primarily to serve as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be utilized as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Deuteration can potentially influence the pharmacokinetic and metabolic characteristics of drugs.