Learn More
Medchemexpress LLC Norfluoxetine (hydrochloride) | 57226-68-3 | 99.6% | 331.76 | 1 MG
SDP
Catalog No. 5000366519
Encompass_Preferred
Norfluoxetine hydrochloride is an active N-demethylated metabolite of Fluoxetine, functioning as a selective serotonin (5-HT) reuptake inhibitor. It is metabolized by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4. This compound also exhibits anticonvulsant activity and inhibits CaV3.3 T current (IC50 = 5 μM).
  • Active N-demethylated metabolite of fluoxetine.
  • Selective serotonin (5-HT) reuptake inhibitor.
  • Metabolized by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4.
  • Inhibits 5-HT uptake.
  • Inhibits CaV3.3 T current (IC50 = 5 μM).
  • Exhibits anticonvulsant activity.
Catalog No. 50-003-66519 Supplier Medchemexpress LLC Supplier No. HYW0112351MG
Norfluoxetine hydrochloride is an active N-demethylated metabolite of Fluoxetine, functioning as a selective serotonin (5-HT) reuptake inhibitor. It is metabolized by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4. This compound also exhibits anticonvulsant activity and inhibits CaV3.3 T current (IC50 = 5 μM).
  • Active N-demethylated metabolite of fluoxetine.
  • Selective serotonin (5-HT) reuptake inhibitor.
  • Metabolized by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4.
  • Inhibits 5-HT uptake.
  • Inhibits CaV3.3 T current (IC50 = 5 μM).
  • Exhibits anticonvulsant activity.