Learn More
Medchemexpress LLC Norfluoxetine hydrochloride | 57226-68-3 | 99.6% | 331.76 | 5 MG
SDP
Catalog No. 5000452845
Encompass_Preferred
Norfluoxetine hydrochloride is an active N-demethylated metabolite of Fluoxetine. Fluoxetine, a selective serotonin (5-HT) reuptake inhibitor, is metabolized into Norfluoxetine hydrochloride by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4.
  • Inhibits 5-HT uptake
  • Inhibits CaV3.3 T current (IC50 = 5 μM)
  • Exhibits anticonvulsant activity
  • Offers protective effect against Pentylenetetrazol-induced epilepsy
Catalog No. 50-004-52845 Supplier Medchemexpress LLC Supplier No. HYW0112355MG
Norfluoxetine hydrochloride is an active N-demethylated metabolite of Fluoxetine. Fluoxetine, a selective serotonin (5-HT) reuptake inhibitor, is metabolized into Norfluoxetine hydrochloride by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4.
  • Inhibits 5-HT uptake
  • Inhibits CaV3.3 T current (IC50 = 5 μM)
  • Exhibits anticonvulsant activity
  • Offers protective effect against Pentylenetetrazol-induced epilepsy