Learn More
Cayman Chemical NorfluoxetInhydrochlorIde 10mg
SDP
Catalog No. 502969564
Encompass_Preferred
An active metabolite of fluoxetine; inhibits 5-HT uptake in rat brain synaptosomal membrane preparations (Ki = 44.7 nM) and isolated human platelets (IC50 = ~15 nM); has been found in the tissues of fish exposed to wastewater effluent
Catalog No. 50-296-9564 Supplier Cayman Chemical Supplier No. 1590010 MG
An active metabolite of fluoxetine; inhibits 5-HT uptake in rat brain synaptosomal membrane preparations (Ki = 44.7 nM) and isolated human platelets (IC50 = ~15 nM); has been found in the tissues of fish exposed to wastewater effluent