Learn More
Norgen Biotek Corp Urine DNA Isolation Micro Kit
Catalog No. NC1541535
Encompass_Preferred
This kit provides a fast, reliable and simple procedure for isolating DNA from up to 1.75 mL of urine. Both high molecular weight DNA (greater than 1 kb in size; mostly cell associated) and the smaller DNA (150 - 250 bp; derived from the circulation) is effectively isolated and purified using a rapid and convenient spin column protocol. Multiple samples can be processed in 30 minutes. This kit can be used to isolate DNA from a broad range of viruses in urine as well. Salts, metabolic wastes, proteins and other contaminants are removed to yield inhibitor-free DNA for use in sensitive applications. The DNA is of excellent quality for various downstream applications such as PCR, qPCR and DNA fingerprinting, methylation studies and more. This kit is fully compatible with Norgen's Urine Collection and Preservation Tubes.
Catalog No. NC1541535 Supplier Norgen Biotek Corp Supplier No. 18100
This kit provides a fast, reliable and simple procedure for isolating DNA from up to 1.75 mL of urine. Both high molecular weight DNA (greater than 1 kb in size; mostly cell associated) and the smaller DNA (150 - 250 bp; derived from the circulation) is effectively isolated and purified using a rapid and convenient spin column protocol. Multiple samples can be processed in 30 minutes. This kit can be used to isolate DNA from a broad range of viruses in urine as well. Salts, metabolic wastes, proteins and other contaminants are removed to yield inhibitor-free DNA for use in sensitive applications. The DNA is of excellent quality for various downstream applications such as PCR, qPCR and DNA fingerprinting, methylation studies and more. This kit is fully compatible with Norgen's Urine Collection and Preservation Tubes.