Learn More
Cayman Chemical Norgestimate, Purity: ≥95%, CAS: 35189-28-7, 10 mg
SDP
Catalog No. 502979601
Encompass_Preferred
A progestin; prodrug form of norelgestromin and levonorgestrel; binds to progestin receptors (mean IC50 = 3.48 nM for rabbit uterine receptors) and is selective for progestin over androgen receptors (mean IC50 = 764 nM for rat prostatic receptors); maintains pregnancy in ovariectomized female rats in a dose-dependent manner; inhibits ovulation in 100% of female rats at 0.5 mg/kg; completely inhibits estrone-induced vaginal cornification in ovariectomized female rats at 2 mg/kg, indicating antiestrogenic activity
Catalog No. 50-297-9601 Supplier Cayman Chemical Supplier No. 1654710 MG
A progestin; prodrug form of norelgestromin and levonorgestrel; binds to progestin receptors (mean IC50 = 3.48 nM for rabbit uterine receptors) and is selective for progestin over androgen receptors (mean IC50 = 764 nM for rat prostatic receptors); maintains pregnancy in ovariectomized female rats in a dose-dependent manner; inhibits ovulation in 100% of female rats at 0.5 mg/kg; completely inhibits estrone-induced vaginal cornification in ovariectomized female rats at 2 mg/kg, indicating antiestrogenic activity