Learn More
Eurofins Discoverx Norgestrel, 10 mg. (92-1135)
Supplier:  Eurofins Discoverx 921135
Encompass
Norgestrel is a synthetic progesterone analog used as an oral contraceptive. It is a racemic mixture of levonorgestrel and dextronorgestrel. Levonorgestrel, the bioactive portion of Norgestrel, inhibits ovulation by causing cervical mucous to thicken. Levonorgestrel has 3 times higher affinity for the progesterone receptor than progesterone. Acts as an agonist for PRα and PRβ. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9667