Learn More
Medchemexpress LLC Norgestrel-d6 | 2376035-98-0 | 99.5% | 5 MG
SDP
Catalog No. 5000405884
Encompass_Preferred
Norgestrel-d6 is the deuterium labeled Levonorgestrel, a synthetic progestogen used as an active ingredient in some hormonal contraceptives. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Levonorgestrel
  • Synthetic progestogen
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • For use as a tracer or internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05884 Supplier Medchemexpress LLC Supplier No. HYB0257S15MG
Norgestrel-d6 is the deuterium labeled Levonorgestrel, a synthetic progestogen used as an active ingredient in some hormonal contraceptives. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Levonorgestrel
  • Synthetic progestogen
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • For use as a tracer or internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS