Learn More
Apexbio Technology LLC Norharmane 244-63-3 1g
SDP
Catalog No. 5000091610
Encompass_Preferred
Norharmane (CAS 244-63-3) is a naturally occurring -carboline alkaloid originally isolated from plants of the Zygophyllaceae family and also present in cooked meats tobacco smoke and coffee Functioning as a co-mutagen norharmane does not display mutagenicity in Salmonella typhimurium TA98 and TA100 assays with or without metabolic activation (S9 mix) but exhibits mutagenic activity in combination with certain non-mutagenic aromatic amines facilitating DNA adduct formation At micromolar concentrations norharmane has been shown to inhibit monoamine oxidase types A and B Norharmane s roles in modulating mutagenesis and enzyme inhibition support its utility in studies of chemical carcinogenesis and neurobiology
Catalog No. 50-000-91610 Supplier Apexbio Technology LLC Supplier No. C49821000
Norharmane (CAS 244-63-3) is a naturally occurring -carboline alkaloid originally isolated from plants of the Zygophyllaceae family and also present in cooked meats tobacco smoke and coffee Functioning as a co-mutagen norharmane does not display mutagenicity in Salmonella typhimurium TA98 and TA100 assays with or without metabolic activation (S9 mix) but exhibits mutagenic activity in combination with certain non-mutagenic aromatic amines facilitating DNA adduct formation At micromolar concentrations norharmane has been shown to inhibit monoamine oxidase types A and B Norharmane s roles in modulating mutagenesis and enzyme inhibition support its utility in studies of chemical carcinogenesis and neurobiology