Learn More
Medchemexpress LLC Norisoboldine | 23599-69-1 | 313.35 | 50 MG
SDP
Catalog No. 5000389666
Encompass_Preferred
Norisoboldine is an orally active natural aryl hydrocarbon receptor (AhR) agonist. It is a major isoquinoline alkaloid found in Radix Linderae and can be used for the research of rheumatoid arthritis and ulcerative colitis.
  • Activates AhR under hypoxic microenvironment.
  • Downregulates mRNA expression of miR-31.
  • Inhibits glycolysis in hypoxia.
  • Promotes Treg differentiation in hypoxia.
  • Facilitates disassociation of HSP90/AhR complexes.
  • Induces nuclear translocation of AhR.
  • Forms AhR/ARNT complexes.
  • Reduces severity of joint swelling and erythema in animal models.
  • Enhances CYP1A1 expression in colons.
  • Suppresses expressions of Glut1 and HK2 in colons.
Catalog No. 50-003-89666 Supplier Medchemexpress LLC Supplier No. HYN058650MG
Norisoboldine is an orally active natural aryl hydrocarbon receptor (AhR) agonist. It is a major isoquinoline alkaloid found in Radix Linderae and can be used for the research of rheumatoid arthritis and ulcerative colitis.
  • Activates AhR under hypoxic microenvironment.
  • Downregulates mRNA expression of miR-31.
  • Inhibits glycolysis in hypoxia.
  • Promotes Treg differentiation in hypoxia.
  • Facilitates disassociation of HSP90/AhR complexes.
  • Induces nuclear translocation of AhR.
  • Forms AhR/ARNT complexes.
  • Reduces severity of joint swelling and erythema in animal models.
  • Enhances CYP1A1 expression in colons.
  • Suppresses expressions of Glut1 and HK2 in colons.