Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Norisoboldine hydrochloride | 5083-84-1 | 99.7% | 349.81 g/mol | C18H20ClNO4 | 10 MG
SDP

Catalog No. 5000297127
Encompass_Preferred

Norisoboldine hydrochloride is an isoquinoline alkaloid salt used as a research chemical and an orally active aryl hydrocarbon receptor (AhR) agonist. It is supplied as a white to off-white solid with high purity and is intended for in vitro and in vivo research applications.

  • high purity (~99.7%)
  • molecular weight 349.81 g/mol
  • CAS number 5083-84-1
  • soluble in DMSO at 100 mg/mL with warming
  • store sealed at -20 °C, protect from moisture
  • available in small research pack sizes (e.g., 10 mg)

Catalog No. 50-002-97127 Supplier Medchemexpress LLC Supplier No. HYN0586A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Norisoboldine hydrochloride is an isoquinoline alkaloid salt used as a research chemical and an orally active aryl hydrocarbon receptor (AhR) agonist. It is supplied as a white to off-white solid with high purity and is intended for in vitro and in vivo research applications.

  • high purity (~99.7%)
  • molecular weight 349.81 g/mol
  • CAS number 5083-84-1
  • soluble in DMSO at 100 mg/mL with warming
  • store sealed at -20 °C, protect from moisture
  • available in small research pack sizes (e.g., 10 mg)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.