Learn More
Cayman Chemical NorketamIn-d4hydrochlorIde 1mg
SDP
Catalog No. 502969568
Encompass_Preferred
An internal standard for the quantification of norketamine by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-296-9568 Supplier Cayman Chemical Supplier No. 158351 MG
An internal standard for the quantification of norketamine by GC- or LC-MS; intended for research and forensic applications