Learn More
Medchemexpress LLC Norleual TFA | 99.9% | 888.97 | 1 MG
SDP
Catalog No. 5000423266
Encompass_Preferred
Norleual TFA is an angiotensin (Ang) IV analog and a hepatocyte growth factor (HGF)/c-Met inhibitor with an IC50 of 3 pM. It acts as an AT4 receptor antagonist and demonstrates potent antiangiogenic activities. It significantly reduces HGF-dependent c-Met and Gab1 phosphorylation and attenuates HGF-dependent c-Met activation and downstream signaling. In vivo, Norleual TFA suppresses pulmonary colonization by B16-F10 murine melanoma cells.
  • Reduces HGF-dependent c-Met and Gab1 phosphorylation
  • Attenuates HGF-dependent c-Met activation and downstream signaling
  • Inhibits HGF-dependent signaling, proliferation, migration, and invasion in multiple cell types
  • Suppresses pulmonary colonization by B16-F10 murine melanoma cells
Catalog No. 50-004-23266 Supplier Medchemexpress LLC Supplier No. HYP1415A1MG
Norleual TFA is an angiotensin (Ang) IV analog and a hepatocyte growth factor (HGF)/c-Met inhibitor with an IC50 of 3 pM. It acts as an AT4 receptor antagonist and demonstrates potent antiangiogenic activities. It significantly reduces HGF-dependent c-Met and Gab1 phosphorylation and attenuates HGF-dependent c-Met activation and downstream signaling. In vivo, Norleual TFA suppresses pulmonary colonization by B16-F10 murine melanoma cells.
  • Reduces HGF-dependent c-Met and Gab1 phosphorylation
  • Attenuates HGF-dependent c-Met activation and downstream signaling
  • Inhibits HGF-dependent signaling, proliferation, migration, and invasion in multiple cell types
  • Suppresses pulmonary colonization by B16-F10 murine melanoma cells