Learn More
Andwin Scientific Norm-Ject Syringes 100cc, 1000/case
SDP
Catalog No. NC9810544
Encompass_Preferred
The Norm-Ject syringes are free of contaminants associated with disposables, latex, rubber, silicone oil, or styrene and are sterile. This product is ideal for sampling, mixing, and filtering. The syringes feature a positive safety stop to avoid accidental spills. This product has accurate and easily readable graduations at 0.5 mL and a volume capacity of 10 mL. It is sold in a case of 1000.
This item is not returnable. View return policy
Catalog No. NC9810544 Supplier Andwin Scientific Supplier No. 53548006
The Norm-Ject syringes are free of contaminants associated with disposables, latex, rubber, silicone oil, or styrene and are sterile. This product is ideal for sampling, mixing, and filtering. The syringes feature a positive safety stop to avoid accidental spills. This product has accurate and easily readable graduations at 0.5 mL and a volume capacity of 10 mL. It is sold in a case of 1000.