Learn More
Carbosynth LLC. Normal Cat Serum, 10 ml
SDP
Catalog No. 501870928
Encompass_Preferred
Normal Cat Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Catalog No. 50-187-0928 Supplier Carbosynth LLC. Supplier No. 88R102610ML
Normal Cat Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2