Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Chicken IgY, Negative Control, R&D Systems™

Catalog No. AB101C
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB101C 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB101C Supplier R&D Systems Supplier No. AB101C
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chicken Polyclonal Antibody has been used in 7 publications

IgY Polyclonal specifically detects IgY in Chicken samples. It is validated for Control.

Specifications

Antigen IgY
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Control
Host Species Chicken
Immunogen NA
Purification Method Ion exchange-purified from egg yolks
Quantity 1 mg
Test Specificity Egg yolks were obtained from naive (non-immunized) chickens and purified for use as normal chicken IgY.
Reconstitution Dissolve the lyophilized chicken IgY in sterile PBS, pH 7.4. If 1 mL of PBS is used, the antibody concentration will be 1 mg/mL.
Target Species Chicken
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.