Learn More
Carbosynth LLC. Normal Dog Serum, 10 ml
SDP
Catalog No. 501870929
Encompass_Preferred
Normal Dog Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Catalog No. 50-187-0929 Supplier Carbosynth LLC. Supplier No. 88R102810ML
Normal Dog Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2