Learn More
Carbosynth LLC. Normal Donkey Serum, 10 ml
SDP
Catalog No. 501870926
Encompass_Preferred
Normal Donkey Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Catalog No. 50-187-0926 Supplier Carbosynth LLC. Supplier No. 88R102210ML
Normal Donkey Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2