Learn More
Abcam Normal Donkey Serum
Catalog No. NC3857548
Encompass_Preferred
Normal Donkey Serum (ab7475) is used for blocking in antibody-based applications e.g. IHC, ICC/IF or as an antibody diluent. It is effective in reducing nonspecific binding thereby maximizing signal-to-noise ratio.- Proven performance: cited in over 265 publications- Available in different sizes to fit your experimental needs- Compatible with our Lab essentials imaging workflow offering.
Catalog No. NC3857548 Supplier Abcam Supplier No. AB7475
Normal Donkey Serum (ab7475) is used for blocking in antibody-based applications e.g. IHC, ICC/IF or as an antibody diluent. It is effective in reducing nonspecific binding thereby maximizing signal-to-noise ratio.- Proven performance: cited in over 265 publications- Available in different sizes to fit your experimental needs- Compatible with our Lab essentials imaging workflow offering.