Learn More
Abcam Normal Donkey Serum, 25ML
Catalog No. NC1697010
Encompass_Preferred
Normal donkey serum is used extensively for the blocking of non-specific antibody binding in tissue and cell staining, and in other applications of antibodies. - Cited in over 190 publications - Suitable for immunocytochemistry/immunofluorescence (ICC/IF), immunohistochemistry (IHC), and more - Superior Fc receptor blocking - Reduces background noise
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1697010 Supplier Abcam Supplier No. AB747525ML
Normal donkey serum is used extensively for the blocking of non-specific antibody binding in tissue and cell staining, and in other applications of antibodies. - Cited in over 190 publications - Suitable for immunocytochemistry/immunofluorescence (ICC/IF), immunohistochemistry (IHC), and more - Superior Fc receptor blocking - Reduces background noise
The product is subject to the following: Abcam Restricted Use Statement