Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Goat Serum (10%)

Catalog No. p-7076119
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8680 15 mL
50-062-Z 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-114-8680 Supplier LIFE TECHNOLOGIES Supplier No. 50197Z
Only null left
Add to Cart
Edge
Add to Cart

Normal Goat Serum (10%)

This 10% Normal Goat Serum blocking solution is ready-to-use, in phosphate buffered saline (pH 7.4) with 0.1% sodium azide. This normal goat serum is intended for use as a blocking solution when a second antibody of goat origin is applied in an immunohistochemical staining procedure.

To block non-specific binding from the goat second antibody, incubate the tissue section with 10% Normal Goat Serum for 10 minutes at room temperature prior to the primary antibody incubation step.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2-8°C
Species Goat/Caprine
Quantity 15 mL
Shipping Condition Wet Ice

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.