Learn More
Alkali Scientific ASI™ Normal Goat Serum, 500mL
SDP
Catalog No. 503029431
Encompass_Preferred
Normal serums are lipid extracted to improve clarity, dialyzed against phosphate buffered saline (PBS) containing sodium azide, and freeze dried. Normal serum diluted to 5% (v/v) in PBS is strongly recommended as a blocking reagent to reduce background from non-specific, conserved sequence and/or Fc receptor binding. Best results are obtained with diluted normal serum from the same host as the labeled antibody, as a separate incubation step before addition of the primary antibody. Virus-Screened Mycoplasma-Tested Triple 0.1 Micron Filtered This product ships on dry ice.
Catalog No. 50-302-9431 Supplier Alkali Scientific Supplier No. FB35
Normal serums are lipid extracted to improve clarity, dialyzed against phosphate buffered saline (PBS) containing sodium azide, and freeze dried. Normal serum diluted to 5% (v/v) in PBS is strongly recommended as a blocking reagent to reduce background from non-specific, conserved sequence and/or Fc receptor binding. Best results are obtained with diluted normal serum from the same host as the labeled antibody, as a separate incubation step before addition of the primary antibody. Virus-Screened Mycoplasma-Tested Triple 0.1 Micron Filtered This product ships on dry ice.