Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Normal Goat Serum

Catalog No. PCN5000
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PCN5000 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PCN5000 Supplier Gibco™ Supplier No. PCN5000
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Normal Goat Serum

Normal goat serum. Whole serum is obtained from non-immunized (preimmune) healthy goats. The serum is clarified through a 0.8 micron low protein binding filter.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Species Goat/Caprine
Form Liquid
Product Type Goat Serum
Quantity 5 mL
Sterility Sterile
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

Analyte Specific Reagent. Analytical and performance characteristics are not established.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.