Learn More
US Biomax Inc NORMAL HEART TISSUE ARRAY,24
SDP
Catalog No. NC2156622 Shop All US Biomax Inc Products
Encompass
Normal heart tissue array, 24 cases/24 cores (1.5mm), replacing HEN241
This item is not returnable. View return policy
Catalog No. NC2156622 Supplier US Biomax Inc Supplier No. HEN241A
Normal heart tissue array, 24 cases/24 cores (1.5mm), replacing HEN241