Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jackson Immuno Research Labs Normal Horse Serum, Unconjugated, 10ml
SDP

Catalog No. NC9909742
Encompass_Preferred

Normal Horse Serum 10.00mL

Catalog No. NC9909742 Supplier Jackson Immuno Research Labs Supplier No. 008000121
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Normal Horse Serum 10.00mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.