Learn More
Quidel Corporation Normal Human Serum Standard Complement 5ml
SDP
Supplier:  Quidel Corporation A113
Encompass
Uniform pool of human serum complement that has been characterized for levels of complement activation fragments as well as CH50.
Catalog No. NC9172398