Learn More
Carbosynth LLC. Normal Human Serum, 10 ml
SDP
Catalog No. 501870938
Encompass_Preferred
Normal Human Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Catalog No. 50-187-0938 Supplier Carbosynth LLC. Supplier No. 90R100210ML
Normal Human Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2