Learn More
Santa Cruz Biotechnology NORMAL MOUSE IGG AGAR CON
Supplier:  Santa Cruz Biotechnology SC2343
Encompass
normal mouse IgG AC,provided at 200 microgram in 0.5 ml volume (125 microliter beads),agarose conjugated normal immunoglobulin, use as negative control for immunoprecipitation
Catalog No. NC9759945