Learn More
Santa Cruz Biotechnology Normal mouse IgG2a, 2 mL
Supplier:  Santa Cruz Biotechnology SC3878
Encompass
Sufficient for 100 tests, non-conjugated, affinity purified immunoglobulin, and for use as negative controls for flow cytometry.
Catalog No. NC9861375