Learn More
Santa Cruz Biotechnology normal mouse IgG2b-PE, 200 µg/ml
Catalog No. NC3733371
Encompass
normal mouse IgG2b-PE is provided at 200 µg in 1.0 ml volumephycoerythrin (PE) conjugated, affinity purified normal immunoglobulin fromrecommended for use as a negative control immunoglobulin in place of a target specific primary antibody of the same isotype (mouse IgG2b) by IF, IHC(P) and FCM applications
Catalog No. NC3733371 Supplier Santa Cruz Biotechnology Supplier No. SC2868
normal mouse IgG2b-PE is provided at 200 µg in 1.0 ml volumephycoerythrin (PE) conjugated, affinity purified normal immunoglobulin fromrecommended for use as a negative control immunoglobulin in place of a target specific primary antibody of the same isotype (mouse IgG2b) by IF, IHC(P) and FCM applications