Learn More
Carbosynth LLC. Normal Pig Serum, 2 ml
SDP
Catalog No. 501870927
Encompass_Preferred
Normal Pig (Swine) Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Catalog No. 50-187-0927 Supplier Carbosynth LLC. Supplier No. 88R10232ML
Normal Pig (Swine) Serum which has been lipid extracted and dialyzed against 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2