Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Normal Porcine Serum

Catalog No. PI31890 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
120 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31890 120 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31890 Supplier Invitrogen™ Supplier No. 31890
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Porcine Control

For blocking purposes, use 5% (v/v) solution (1:20 dilution from rehydrated volume). Rehydration according to instruction yields 100% serum. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, freeze rehydrated undiluted product in small aliquots at 20°C. Country of Origin: USA.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Lyophilized
Product Type Normal Porcine Serum
Quantity 120 mg
Concentration 60.0 mg/mL
Formulation Whole serum, PBS with 0.05% sodium azide; pH 7.6
For Use With (Application) Immunohistochemistry, Immunocytochemistry, Control, Blocking Assay
Isotype Ig
The antibody I purchased from you says ''concentration not determined'' on the product label and on the product page. How do I determine the concentration?

Some antibodies are provided in non-purified formats, such as ascites fluid, tissue culture supernatant, or whole serum. These formats contain multiple proteins, which can interfere with accurate protein quantification. Traditional methods like A280 measurement or protein assays such as BCA quantify total protein but do not provide a specific concentration for the antibody in question.

Literature, such as "Antibodies" by Harlow & Lane (1988) (https://antibodiesmanual.org/index.php) (Table 8.2 - Storing and Purifying Antibodies), suggests approximate concentrations of antibodies in these solutions, as shown below. However, please note that these are rough estimates, and actual concentrations can vary based on several factors.

Serum
Total antibody concentration: 10 mg/mL
Specific antibody concentration: 0.1 to 1 mg/mL (a maximum of 10% of the antibody population is expected to be the specific antibody of interest)

Tissue Culture Supernatant (10% FBS)
Total antibody concentration: 1 mg/mL
Specific antibody concentration: 0.05 mg/mL

Tissue Culture Supernatant (serum-free)
Total antibody concentration: 0.01 to 0.05 mg/mL
Specific antibody concentration: 0.01 to 0.05 mg/mL

Ascites
Total antibody concentration: 1 to 10 mg/mL
Specific antibody concentration: 0.9 to 9 mg/mL

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.